Skip to Content
Merck
All Photos(2)

Key Documents

850699P

Avanti

08:0 PE

1,2-dioctanoyl-sn-glycero-3-phosphoethanolamine, powder

Synonym(s):

PE(8:0/8:0)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€227.00
25 MG
€379.00

€227.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€227.00
25 MG
€379.00

About This Item

Empirical Formula (Hill Notation):
C21H42NO8P
CAS Number:
Molecular Weight:
467.53
UNSPSC Code:
51191904
NACRES:
NA.25

€227.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (850699P-10mg)
pkg of 1 × 25 mg (850699P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850699P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC)=O)COC(CCCCCCC)=O

InChI key

UNACBKDVIYEXSL-LJQANCHMSA-N

General description

Phosphatidylethanolamine (PE) is a glycerophospholipid.[1] This nonbilayer-forming phospholipid has a cone shape due to the presence of its head group.[2]

Application

08:0 PE (1,2-dioctanoyl-sn-glycero-3-phosphoethanolamine) may be used in the preparation of DOPC liposomes for pH sensor conjugation in imaging pH changes in living cells.[3]

Biochem/physiol Actions

Phosphatidylethanolamine (PE) plays a vital role in the structure and function of eukaryotic membranes.[1] It serves as a lipid chaperon. It is known to participate in the initiation of autophagy. The hexagonal phase of PE is involved in membrane fusion events.[2]

Packaging

5 mL Clear Glass Sealed Ampule (850699P-10mg)
5 mL Clear Glass Sealed Ampule (850699P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jungju Yoon et al.
Advanced materials (Deerfield Beach, Fla.), 26(26), 4559-4564 (2014-05-03)
Phase separation in films of phospholipids and conjugated polymers results in nanoassemblies because of a difference in the physicochemical properties between the hydrophobic polymers and the polar lipid heads, together with the comparable polymer side-chain lengths to lipid tail lengths
Ronja Marie Kühnel et al.
The Analyst, 144(9), 3030-3037 (2019-03-23)
The design of ion sensors has gained importance for the study of ion dynamics in cells, with fluorescent proton nanosensors attracting particular interest because of their applicability in monitoring pH gradients in biological microcompartments and reconstituted membrane systems. In this
Federica Gibellini et al.
IUBMB life, 62(6), 414-428 (2010-05-27)
The glycerophospholipids phosphatidylcholine (PC) and phosphatidylethanolamine (PE) account for greater than 50% of the total phospholipid species in eukaryotic membranes and thus play major roles in the structure and function of those membranes. In most eukaryotic cells, PC and PE
Dhaval Patel et al.
Oxidative medicine and cellular longevity, 2017, 4829180-4829180 (2017-08-09)
Phosphatidylethanolamine (PE) is the second most abundant phospholipid in mammalian cells. PE comprises about 15-25% of the total lipid in mammalian cells; it is enriched in the inner leaflet of membranes, and it is especially abundant in the inner mitochondrial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service