Skip to Content
Merck
All Photos(2)

Key Documents

850556O

Avanti

Phytantriol (mixed isomers)

Avanti Research - A Croda Brand 850556O

Synonym(s):

3,7,11,15-tetramethylhexadecane-1,2,3-triol (mixed isomers)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€286.00

€286.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€286.00

About This Item

Empirical Formula (Hill Notation):
C20H42O3
CAS Number:
Molecular Weight:
330.55
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€286.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 g (850556O-1g)

manufacturer/tradename

Avanti Research - A Croda Brand 850556O

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(C)CCCC(C)CCCC(C)CCCC(C)(O)C(O)CO

InChI

1S/C20H42O3/c1-16(2)9-6-10-17(3)11-7-12-18(4)13-8-14-20(5,23)19(22)15-21/h16-19,21-23H,6-15H2,1-5H3

InChI key

CGIHFIDULQUVJG-UHFFFAOYSA-N

General description

The temperature−composition phase diagram of phytantriol/water mixtures is qualitatively similar to that of aqueous glycerol monooleate so its stability makes it an interesting alternative to exploit various applications of the cubic liquid crystalline phases.

Biochem/physiol Actions

Phytantriol is a majorly used ingredient in skin and hair based cosmetics. It is viscous and enhances penetration of amino acids and vitamin components of cosmetics.[1] It is structurally stable than glyceryl monooleate (GMO). Phytantriol in combination with the polymer (F127) and a hydrotrope (propyleneglycol) is used in the preparation of cubosomes for protein or vaccine delivery.[2]

Packaging

60 mL Amber Wide Mouth Screw Cap Glass Bottle (850556O-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phase behavior of the phytantriol/water system
Barauskas J and Landh T
Langmuir, 19(23), 9562-9565 (2003)
S B Rizwan et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 79(1), 15-22 (2011-01-18)
Different delivery strategies to improve the immunogenicity of peptide/protein-based vaccines are currently under investigation. In this study, the preparation and physicochemical characterisation of cubosomes, a novel lipid-based particulate system currently being explored for vaccine delivery, was investigated. Cubosomes were prepared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service