Skip to Content
Merck
All Photos(2)

Key Documents

850479P

Avanti

16:0-18:0(4,5-dibromo) PC

Avanti Research - A Croda Brand 850479P, powder

Synonym(s):

1-palmitoyl-2-stearoyl(4,5)dibromo-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H82Br2NO8P
CAS Number:
Molecular Weight:
919.88
UNSPSC Code:
12352211

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (850479P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850479P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCC(Br)C(Br)CCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O

General description

16:0-18:0(4,5-dibromo) PC
(1-palmitoyl-2-stearoyl (4,5) dibromo-sn-glycero-3-phosphocholine) is a lipid which has a phosphocholine backbone, palmitoyl and stearoyl fatty acid chains. The 4th and 5th carbons of stearoyl group has bromo substitution.

Application

16:0-18:0(4,5-dibromo) PC (1-palmitoyl-2-stearoyl (4,5) dibromo-sn-glycero-3-phosphocholine) may be used as a component in the vesicles for quenching experiments.[1]

Biochem/physiol Actions

16:0-18:0(4,5-dibromo) PC (1-palmitoyl-2-stearoyl (4,5) dibromo-sn-glycero-3-phosphocholine) can be used in vesicle preparation.[1]

Packaging

5 mL Clear Glass Sealed Ampule (850479P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cari M Anderson et al.
The journal of physical chemistry. B, 123(1), 170-179 (2018-11-28)
The interactions between small molecules and lipid bilayers play a critical role in the function of cellular membranes. Understanding how a small molecule interacts with the lipid bilayer differently based on its charge reveals primordial mechanisms of transport across membranes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service