Skip to Content
Merck
All Photos(2)

Key Documents

850461C

Avanti

16:0-22:6 PC

Avanti Research - A Croda Brand

Synonym(s):

1-hexadecanoyl-2-(4Z,7Z,10Z,13Z,16Z,19Z-docosahexaenoyl)-sn-glycero-3-phosphocholine; PC(16:0/22:6(4Z,7Z,10Z,13Z,16Z,19Z))

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€295.00

€295.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€295.00

About This Item

Empirical Formula (Hill Notation):
C46H80NO8P
CAS Number:
Molecular Weight:
806.10
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€295.00


Please contact Customer Service for Availability

Request a Bulk Order

description

1-palmitoyl-2-docosahexaenoyl-sn-glycero-3-phosphocholine, chloroform

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850461C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (850461C-25mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](OC(=O)CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CC)COC(=O)CCCCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C46H80NO8P/c1-6-8-10-12-14-16-18-20-21-22-23-24-25-27-29-31-33-35-37-39-46(49)55-44(43-54-56(50,51)53-41-40-47(3,4)5)42-52-45(48)38-36-34-32-30-28-26-19-17-15-13-11-9-7-2/h8,10,14,16,20-21,23-24,27,29,33,35,44H,6-7,9,11-13,15,17-19,22,25-26,28,30-32,34,36-43H2,1-5H3/b10-8-,16-14-,21-20-,24-23-,29-27-,35-33-/t44-/m1/s1

InChI key

IESVDEZGAHUQJU-ZLBXKVHBSA-N

General description

16:0-22:6 PC or 1-palmitoyl-2-docosahexaenoyl-sn-glycero-3-phosphocholine is a glycerophospholipid, with substitution of palmitic acid and docosahexaenic acid at sn-1 and sn-2 positions of the glycerol backbone, respectively.

Application

16:0-22:6 PC or 1-palmitoyl-2-docosahexaenoyl-sn-glycero-3-phosphocholine:
  • has been used in vesicle preparation[1]
  • has been used in liposome extrusion and liposome flotation assay[2]
  • might be used in large unilamellar vesicles (LUVs) to develop a scattering density profile (SDP) model for lipids with polyunsaturated fatty acid (PUFA)[3]

Packaging

5 mL Clear Glass Sealed Ampule (850461C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A universal method for planar lipid bilayer formation by freeze and thaw
Sugihara K, et al.
Soft Matter, 8(20), 5525-5531 (2012)
Anddre Osmar Valdivia et al.
ACS omega, 5(25), 15454-15467 (2020-07-09)
Multiple sclerosis has complex pathogenesis encompassing a variety of components (immunologic, genetic, and environmental). The autoimmunogenicity against the host's myelin basic protein is a major contributor. An increase in myelin basic protein deimination (a post-translational modification) and a change in
Drew Marquardt et al.
Chemistry and physics of lipids, 229, 104892-104892 (2020-02-18)
We present the detailed structural analysis of polyunsaturated fatty acid-containing phospholipids namely, 1-palmitoyl-2-docosahexaenoyl-sn-glycero-3-phosphocholine (PDPC) and 1-stearoyl-2-docosahexaenoyl-sn-glycero-3-phosphocholine (SDPC). A newly developed molecular dynamics (MD) simulation parsing scheme for lipids containing fatty acids with multiple double bonds was implemented into the scattering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service