Skip to Content
Merck
All Photos(2)

Key Documents

850340C

Avanti

13:0 PC

1,2-ditridecanoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

PC(13:0/13:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€118.00

€118.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€118.00

About This Item

Empirical Formula (Hill Notation):
C34H68NO8P
CAS Number:
Molecular Weight:
649.88
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€118.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850340C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850340C

concentration

10 mg/mL (850340C-25mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

General description

13:0 PC is ditridecanoyl phosphatidyl choline (DTPC).[1] Phosphatidylcholine (PC) is a phospholipid, present in endothelial cells, corneal epithelial cells, and fibroblast.[2] Structurally, PC has a polar head consisting of choline and phosphate and two non-polar tails consisting of fatty acids. The polar head group is water soluble and the non-polar tails are water insoluble. Due to its amphiphilic properties, it is present in the cell membrane. In the human body, liver synthesizes PC. Dietary sources of PC are egg and soybeans.[3]
Phosphatidylcholine (PC), a phospholipid carries a positively charged quaternary amine and a negatively charged phosphate in its headgroup. It can form strong bilayer and is an important component of mammalian membranes.[4]
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

13:0 PC (1,2-ditridecanoyl-sn-glycero-3-phosphocholine) may be used as a lipid standard for the quantification of phosphatidylcholines (PCs) by mass spectrometric analysis.[5][6][6]
13:0 Phosphatidylcholine (PC) has been used as a lipid standard for mass spectrometry.[7][2][8]

Biochem/physiol Actions

Phosphatidylcholine (PC) is the precursor for the triacylglycerol synthesis. The concentration of PC decreases owing to enhanced triacylglycerol production in the liver. Phosphatidylcholine may be a biomarker for the diagnosis of fatty liver.[7] Its structure enables them to be used in food industry as emulsifiers.[3]
Phosphatidylcholine (PC) can serve as a surfactant in the mucus.[9]

Packaging

5 mL Clear Glass Sealed Ampule (850340C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genea Edwards et al.
Biochimie, 101, 232-247 (2014-02-25)
To compare phospholipid (phosphatidylcholine, phosphatidylserine, phosphatidylethanolamine and phosphatidylinositol) profiles of human control and glaucomatous aqueous humor (AQH). AQH samples were procured during surgery from human POAG and control subjects (n = 15 each). Samples were used following institutional review board
Phospholipid makeup of the breast adipose tissue is impacted by obesity and mammary cancer in the mouse: Results of a pilot study
Margolis M, et al.
Biochimie, 108(11), 133-139 (2015)
Lipid-Protein Interactions in Membranes
The Membranes of Cells, 291-334 null
Inflammatory Bowel Disease
Integrative Medicine, 501-516 null
Julie M Davies et al.
PloS one, 9(12), e114352-e114352 (2014-12-04)
Intake of saturated fat is a risk factor for ulcerative colitis (UC) and colon cancer. Changes in the microbiota have been implicated in the development of UC and colon cancer. The host and the microbiota generate metabolites that may contribute

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service