Skip to Content
Merck
All Photos(2)

Documents

850184P

Avanti

08:0 PI(3,5)P2

1,2-dioctanoyl-sn-glycero-3-phospho-(1′-myo-inositol-3′,5′-bisphosphate) (ammonium salt), powder

Synonym(s):

PIP2[3′,5′](8:0/8:0); 110664

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H58N3O19P3
CAS Number:
Molecular Weight:
797.66
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (850184P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (850184P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand 850184P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](OP(O)([O-])=O)[C@H](O)[C@@H](OP([O-])(O)=O)[C@H]1O)=O)(OC(CCCCCCC)=O)COC(CCCCCCC)=O.[NH4+].[NH4+].[NH4+]

General description

Inositol phospholipid species are membrane-bound signaling molecules that have been implicated in almost all aspects of cellular physiology, including cellular growth, metabolism, proliferation, and survival.
Phosphatidylinositol 3,5-bisphosphate (PI(3,5)P2) is the least found phosphoinositides (PIs) and it accounts for approximately 0.05 to 0.1% of total cellular PI.

Biochem/physiol Actions

Phosphatidylinositol 3,5-bisphosphate (PI(3,5)P2) plays a key role in the embryonic development. It is very essential at postnatal stages. It is capable of activating ion channels on endosomes and lysosomes.

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850184P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850184P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junya Hasegawa et al.
Cell structure and function, 42(1), 49-60 (2017-03-18)
In most eukaryotes, phosphoinositides (PIs) have crucial roles in multiple cellular functions. Although the cellular levels of phosphatidylinositol 5-phosphate (PI5P) and phosphatidylinositol 3,5-bisphosphate (PI(3,5)P2) are extremely low relative to some other PIs, emerging evidence demonstrates that both lipids are crucial
Bong-Kwan Han et al.
The Journal of biological chemistry, 288(28), 20633-20645 (2013-06-05)
Glucose/carbon metabolism is a fundamental cellular process in living cells. In response to varying environments, eukaryotic cells reprogram their glucose/carbon metabolism between aerobic or anaerobic glycolysis, oxidative phosphorylation, and/or gluconeogenesis. The distinct type of glucose/carbon metabolism that a cell carries
Gerald R V Hammond et al.
Science (New York, N.Y.), 337(6095), 727-730 (2012-06-23)
The quantitatively minor phospholipid phosphatidylinositol (4,5)-bisphosphate [PI(4,5)P(2)] fulfills many cellular functions in the plasma membrane (PM), whereas its synthetic precursor, phosphatidylinositol 4-phosphate (PI4P), has no assigned PM roles apart from PI(4,5)P(2) synthesis. We used a combination of pharmacological and chemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service