Skip to Content
Merck
All Photos(2)

Key Documents

840886C

Avanti

20:4 PA

1,2-diarachidonoyl-sn-glycero-3-phosphate (sodium salt), chloroform

Synonym(s):

1,2-di-(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-phosphate (sodium salt); DAPA; PA(20:4(5Z,8Z,11Z,14Z)/20:4(5Z,8Z,11Z,14Z))

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€612.00

€612.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€612.00

About This Item

Empirical Formula (Hill Notation):
C43H68O8PNa
CAS Number:
Molecular Weight:
766.96
UNSPSC Code:
51191904
NACRES:
NA.25

€612.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840886C-25mg)
pkg of 2 × 4 mL (840886C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840886C

concentration

10 mg/mL (840886C-25mg)
25 mg/mL (840886C-200mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O)=O)(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)COC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O.[Na+]

InChI

1S/C43H69O8P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-42(44)49-39-41(40-50-52(46,47)48)51-43(45)38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h11-14,17-20,23-26,29-32,41H,3-10,15-16,21-22,27-28,33-40H2,1-2H3,(H2,46,47,48);/q;+1/p-1/b13-11-,14-12-,19-17-,20-18-,25-23-,26-24-,31-29-,32-30-;/t41-;/m1./s1

InChI key

FDDKKAHLSUVCLC-QHEFBXHFSA-M

General description

Phosphatidic acid (PA) is a precursor of lysophosphatidic acid (LPA) and is present in leafy vegetables.[1] 20:4 PA or 1,2-diarachidonoyl-sn-glycero-3-phosphate (sodium salt) is a sodium salt of phosphatidic acid, with substitution of two chains of arachidonic acid at the sn-1 and sn-2 positions, in the glycerol backbone.

Application

20:4 PA or 1,2-diarachidonoyl-sn-glycero-3-phosphate (sodium salt) has been used as a supplement in mouse chow to see changes in small intestine gene expression.[1]

Packaging

5 mL Clear Glass Sealed Ampule (840886C-200mg)
5 mL Clear Glass Sealed Ampule (840886C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamad Navab et al.
Journal of lipid research, 54(12), 3403-3418 (2013-10-03)
We recently reported that levels of unsaturated lysophosphatidic acid (LPA) in the small intestine significantly correlated with the extent of aortic atherosclerosis in LDL receptor-null (LDLR⁻/⁻) mice fed a Western diet (WD). Here we demonstrate that WD increases unsaturated (but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service