Skip to Content
Merck
All Photos(2)

Documents

840513P

Avanti

18:1 Phosphatidylethanol

1,2-dioleoyl-sn-glycero-3-phosphoethanol (sodium salt), powder

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-phosphoethanol (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H76O8PNa
CAS Number:
Molecular Weight:
751.00
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (840513P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840513P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C41H77O8P.Na/c1-4-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-40(42)46-37-39(38-48-50(44,45)47-6-3)49-41(43)36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-5-2;/h19-22,39H,4-18,23-38H2,1-3H3,(H,44,45);/q;+1/p-1/b21-19-,22-20-;/t39-;/m1./s1

InChI key

AHVVHDCFEFPKMN-AOEFFCPUSA-M

General description

18:1 Phosphatidylethanol (PEth)/1,2-dioleoyl-sn-glycero-3-phosphoethanol is a sodium salt of modified lipid, wherein C-1, C-2 of glycerol is substituted with 9Z-octadecenolyl and C-3 of glycerol is substituted by phosphoethanol group. This lipid metabolite of ethanol has two oleic acid chains.

Application

18:1 Phosphatidylethanol (PEth)/1,2-dioleoyl-sn-glycero-3-phosphoethanol may be used:
  • as a standard in liquid chromatography−mass spectrometry (LC-MS) analysis of PEth in human whole blood
  • as a reference material in quantifying phosphatidylethanol in human blood by liquid chromatography high resolution mass spectrometry (LC−HRMS )
  • to incorporate into human high density lipoprotein (HDL) particles to study its effects on the secretion of vascular endothelial growth factor (VEGF) in cultured human smooth muscle cells

Biochem/physiol Actions

Phosphatidylethanol (PEth) obtained from phosphatidylcholine in erythrocytes can acts as an important biomarker for alcohol consumption.

Packaging

5 mL Clear Glass Sealed Ampule (840513P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marja K Liisanantti et al.
Atherosclerosis, 180(2), 263-269 (2005-05-25)
To study whether qualitative changes in high density lipoprotein (HDL) phospholipids mediate part of the advantageous effects of ethanol on atherosclerosis, we investigated whether HDL associated phosphatidylethanol (PEth) affects the secretion of vascular endothelial growth factor (VEGF) from cultured human
Xiaoyu Liang et al.
Molecular psychiatry (2020-02-09)
The process of diagnosing hazardous alcohol drinking (HAD) is based on self-reported data and is thereby vulnerable to bias. There has been an interest in developing epigenetic biomarkers for HAD that might complement clinical assessment. Because alcohol consumption has been
Alessandro Nalesso et al.
Journal of chromatography. A, 1218(46), 8423-8431 (2011-10-18)
Phosphatidylethanol (PEth) is a group of aberrant phospholipids formed in cell membranes in the presence of ethanol by the catalytic action of the enzyme phospholipase D on phosphatidylcholine. Recently published literature has demonstrated the existence of several molecular species of
Siming Wang et al.
Talanta, 166, 315-320 (2017-02-19)
Phosphatidylethanol (PEth) is a special phospholipid that is only formed in the presence of ethanol, and therefore, serves as a promising biomarker for alcohol intake. In this study, a simple, rapid and precise method based on LC-MS/MS combined with ultrasound-assisted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service