Skip to Content
Merck
All Photos(2)

Key Documents

840446P

Avanti

15:0 PG

1,2-dipentadecanoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), powder

Synonym(s):

PG(15:0/15:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€183.00

€183.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€183.00

About This Item

Empirical Formula (Hill Notation):
C36H70O10PNa
CAS Number:
Molecular Weight:
716.90
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€183.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (840446P-25mg)
pkg of 1 × 500 mg (840446P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840446P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCCC)OCC(O)CO

InChI

1S/C36H71O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-35(39)43-31-34(32-45-47(41,42)44-30-33(38)29-37)46-36(40)28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h33-34,37-38H,3-32H2,1-2H3,(H,41,42);/q;+1/p-1/t33?,34-;/m1./s1

InChI key

SWGQXCDMOFBUIE-XWYVMIOLSA-M

General description

Phosphatidylglycerol (PG) is an anionic phospholipid. It is present in the membranes of mitochondria and microsomes. It is an important constituent of the lung surfactant. In lungs, PG is mainly located in the lamella body membranes.[1]

Application

15:0 PG (1,2-dipentadecanoyl-sn-glycero-3-phospho-(1′-rac-glycerol)) has been used as an internal standard for the polar lipids (PLs) fraction in fatty acid analysis[2][3] and biomass composition analyses by gas chromatography–flame ionization detection (GC–FID).[4]

Biochem/physiol Actions

Phosphatidylglycerol (PG) can serve as a protein kinase C family activator.[1]

Packaging

20 mL Clear Glass Screw Cap Vial (840446P-500mg)
5 mL Amber Glass Screw Cap Vial (840446P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of nitrogen addition on lipid productivity of nitrogen starved Nannochloropsis gaditana
Janssen JH, et al.
Algal research (2018)
Effect of initial biomass-specific photon supply rate on fatty acid accumulation in nitrogen depleted Nannochloropsis gaditana under simulated outdoor light conditions
Janssen JH, et al.
Algal research (2018)
Translocation and de novo synthesis of eicosapentaenoic acid (EPA) during nitrogen starvation in Nannochloropsis gaditana
Janssen JH, et al.
Algal research (2019)
Shin-Ya Morita et al.
Scientific reports, 5, 11737-11737 (2015-07-01)
Phosphatidylglycerol (PG) and cardiolipin (CL) are synthesized in mitochondria and regulate numerous biological functions. In this study, a novel fluorometric method was developed for measuring PG and CL using combinations of specific enzymes and Amplex Red. This assay quantified the
Rose M Gathungu et al.
Analytical chemistry, 90(22), 13523-13532 (2018-09-29)
Lipidomics requires the accurate annotation of lipids in complex samples to enable determination of their biological relevance. We demonstrate that unintentional in-source fragmentation (ISF, common in lipidomics) generates ions that have identical masses to other lipids. Lysophosphatidylcholines (LPC), for example

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service