Skip to Content
Merck
All Photos(1)

Key Documents

810600C

Avanti

16:0-12 Doxyl PC

Avanti Research - A Croda Brand 810600C

Synonym(s):

1-palmitoyl-2-stearoyl-(12-doxyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H90N2O10P
CAS Number:
Molecular Weight:
862.19
UNSPSC Code:
51321705
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810600C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810600C

concentration

1 mg/mL (810600C-1mg)

lipid type

phospholipids
ESR probes

shipped in

dry ice

storage temp.

−20°C

General description

Avanti′s nitroxide spin product listing is a group of compounds designed to act as membrane probes. A variety of positions down the hydrophobic chain are labeled with the nitroxide functional groups to allow probing the membrane at various depths. These compounds have been synthesized from 1-palmitoyl-2-hydroxy-sn-glycerol-3-phosphocholine with the product being purified by column chromatography. Various n-doxyl phosphocholines have been recently used as biophysical tools to elucidate membrane trafficking with phosphatidylinositol transfer proteins[1] and as fluorescent quenchers in lipid bilayer structural studies.[2]

Packaging

5 mL Clear Glass Sealed Ampule (810600C-1mg)

Preparation Note

To prevent aggregation, prepare water-based solutions of 2 mM stock solutions of n-DOXYL PCs and store in plastic. Dilute stock solutions to 0.03- 0.1 mM solutions for EPR studies.[3] For liposome preparations in fluorescent quenching measurements, dissolve the doxyl lipid in 150 μl absolute ethanol for a concentration of 40.3 mM.[2],Supplemental information

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ling Yan et al.
FEBS letters, 555(3), 545-550 (2003-12-17)
We investigated the ability of tBid (truncated form of Bid) to bind and permeabilize the liposomes (large unilamellar vesicles, LUVs) and release fluorescent marker molecules (fluorescein-isothiocyanate-conjugated dextrans, FITC-dextrans) of various molecular diameters (FD-20, FD-70, FD-250S) from LUVs. Obtained data showed
Kervin O Evans et al.
Biochimica et biophysica acta, 1848(5), 1175-1182 (2015-02-18)
The phenols hydroxytyrosol and tyrosol made abundantly available through olive oil processing were enzymatically transesterified into effective lipophilic antioxidants with cuphea oil. The hydroxytyrosyl and tyrosyl esters made from cuphea oil were assessed for their ability to partition into, locate
L A Falls et al.
The Journal of biological chemistry, 276(26), 23895-23902 (2001-04-20)
The hydrophobic omega-loop within the prothrombin gamma-carboxyglutamic acid-rich (Gla) domain is important in membrane binding. The role of this region in membrane binding was investigated using a synthetic peptide, PT-(1-46)F4W, which includes the N-terminal 46 residues of human prothrombin with
Bhagyashree D Rao et al.
Chemistry and physics of lipids, 226, 104849-104849 (2019-12-15)
pH (low) insertion peptide (pHLIP) is a polypeptide from the third transmembrane helix of bacteriorhodopsin. The pH-dependent membrane insertion of pHLIP has been conveniently exploited for translocation of cargo molecules and as a novel imaging agent in cancer biology due
Local polarity and hydrogen bonding inside the Sec14p phospholipid-binding cavity: high-field multi-frequency electron paramagnetic resonance studies.
Smirnova TI, et al.
Biophysical Journal, 92, 3686-3695 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service