Skip to Content
Merck
All Photos(1)

Key Documents

810383C

Avanti

18:0 PE-TopFluor AF488

Avanti Research - A Croda Brand

Synonym(s):

1,2-Distearyl-sn-glycero-3-phosphoethanolamine-N-( TopFluor AF488) ammonium salt

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
€276.00

€276.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 μG
€276.00

About This Item

Empirical Formula (Hill Notation):
C62H103N6O18PS2
CAS Number:
Molecular Weight:
1315.61
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€276.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

chloroform solution

packaging

package of 1 × 250 μg

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

250 μg/mL in chloroform

shipped in

dry ice

storage temp.

−20°C

Packaging

5 mL Amber Glass Screw Cap Vial (810383C-250UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service