Skip to Content
Merck
All Photos(2)

Key Documents

810265P

Avanti

C11 TopFluor Sphingomyelin

N-[11-(dipyrrometheneboron difluoride)undecanoyl]-D-erythro-sphingosylphosphorylcholine, powder

Synonym(s):

N-[11-(TopFluor)undecanoyl]-D-erythro-sphingosine-1-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
€390.00

€390.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 μG
€390.00

About This Item

Empirical Formula (Hill Notation):
C47H82N4O6PF2
CAS Number:
Molecular Weight:
868.15
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€390.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 250 μg (810265P-250ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810265P

shipped in

dry ice

storage temp.

−20°C

General description

Sphingomyelin is a membrane phospholipid with a central sphingosine and an amide linked fatty acid. Phosphocholine forms the polar head group.[1] It is one of the major phospholipid present in milk fat globule membrane.[2]

Biochem/physiol Actions

Sphingomyelin controls cell growth, differentiation, apoptosis and signal transduction along the membrane. It is known to prevent the proliferation of colon tumour. Sphingomyelin also inhibits cholesterol absorption.[2] Abnormal accumulation of sphingomyelin causes Niemann Pick disease.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (810265P-250ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Apoptosis: Biology and Mechanisms, 66-66 (2013)
Bioactive Components of Milk, 22-22 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service