Skip to Content
Merck
All Photos(1)

Key Documents

810242P

Avanti

TopFluor TMR PS

Avanti Research - A Croda Brand 810242P, powder

Synonym(s):

1-oleoyl-2-(6-((4,4-difluoro-1,3-dimethyl-5-(4-methoxyphenyl)-4-bora-3α,4α-diaza-s-indacene-2-propionyl)amino)hexanoyl)-sn-glycero-3- phospho-L-serine (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€652.00

€652.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€652.00

About This Item

Empirical Formula (Hill Notation):
C51H79 BF2N5 O12P
CAS Number:
Molecular Weight:
1033.98
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€652.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810242P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810242P

shipped in

dry ice

storage temp.

−20°C

General description

TopFluor TMR PS is an acyl-chain-modified fluorescent phospholipid.[1] Phosphotidylserine is considered as a vital lipid and is found in the inner mitochondrial region.[2]

Application

TopFluor TMR PS is suitable for the formation of lipid membranes[1] in microsystem.[3]

Biochem/physiol Actions

TopFluor TMR PS might be useful to enclose specific molecules into micro-reactors for the formation of concentration gradient.[4] It is also useful for cellular labelling.

Packaging

5 mL Clear Glass Sealed Ampule (810242P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single-molecule analysis of phospholipid scrambling by TMEM16F
Watanabe R, et al.
Proceedings of the National Academy of Sciences of the USA (2018)
PS (PhosphatidylSerine) Nature's Brain Booster A Vital Lipid Nutrient For Memory Mood and Stress, 84-84 (2005)
High-throughput single-molecule bioassay using micro-reactor arrays with a concentration gradient of target molecules
Watanabe R, et al.
Lab on a chip, 18(18), 2849-2853 (2018)
Microsystem for the single molecule analysis of membrane transport proteins
Watanabe R
Biochim. Biophys. Acta Gen. Subj. (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service