Skip to Content
Merck
All Photos(2)

Key Documents

810162C

Avanti

14:0-12:0 NBD PG

Avanti Research - A Croda Brand 810162C

Synonym(s):

1-myristoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-[phospho-rac-(1-glycerol)] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H68N5O13P
CAS Number:
Molecular Weight:
833.95
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810162C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810162C

concentration

1 mg/mL (810162C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

NBD PG is a lipid fluorescent probe with 7-nitro-2-1,3-benzoxadiazol-4-yl (NBD) attached to phosphatidylglycerol (PG). PG is localized to alveoli and is mostly found in the pulmonary surfactant.

Biochem/physiol Actions

7-nitro-2-1,3-benzoxadiazol-4-yl (NBD) labeled fluorescence probes efficiently contributes to study the formation of membranes, micelles and reverse micelles. 7-nitro-2-1,3-benzoxadiazol-4-yl (NBD) phosphatidylglycerol (PG) serves as a substrate for enzyme assays. PG plays a vital role in the regulation of innate immune response and contributes to alveolar stability.

Packaging

5 mL Amber Glass Screw Cap Vial (810162C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service