Skip to Content
Merck
All Photos(1)

Key Documents

810110P

Avanti

12-NBD Stearate

Avanti Research - A Croda Brand 810110P, powder

Synonym(s):

12-N-methyl-(7-nitrobenz-2-oxa-1,3-diazo)aminostearic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H40N4O5
CAS Number:
Molecular Weight:
476.61
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810110P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810110P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[N+](=O)([O-])c1c2n[o]nc2c(cc1)N(C(CCCCCCCCCCC(=O)O)CCCCCC)C

InChI

1S/C25H40N4O5/c1-3-4-5-12-15-20(16-13-10-8-6-7-9-11-14-17-23(30)31)28(2)21-18-19-22(29(32)33)25-24(21)26-34-27-25/h18-20H,3-17H2,1-2H3,(H,30,31)

InChI key

SAAVELCIPHKGJE-UHFFFAOYSA-N

General description

12-NBD Stearate is amphipathic, nonesterifiable fluorescent fatty acid probe and is a long-chain fatty acid (LCFA) derivative.

Application

12-NBD Stearate has been used in the fluorescent labeling of omega-3 fatty acid in biomaterial films and as a ligand for fatty acid binding protein 5 (FABP5) in competitive binding based high throughput fluorescence displacement assay. It may be used a ligand fluorescence displacement assay of fatty-acid binding protein 5 (FABP5).

Biochem/physiol Actions

12-NBD Stearate incorporated in lipid droplets and is used majorly in fluorescence displacement binding studies of fatty-acid binding proteins. The sterol carrier protein-2 (SCP-2) 15-kDa isoform favours NBD Stearate transport. The uptake of NBD Stearate is regulated by fatty acid binding protein (FABP) expression regulates uptake and inhibited by filipin III.

Packaging

5 mL Amber Glass Screw Cap Vial (810110P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lu Bai et al.
ACS infectious diseases, 6(4), 637-648 (2020-02-14)
Assembly of the bacterial cell wall requires not only the biosynthesis of cell wall components but also the transport of these metabolites to the cell exterior for assembly into polymers and membranes required for bacterial viability and virulence. LprG is
Targeting fatty acid binding protein (FABP) anandamide transporters-a novel strategy for development of anti-inflammatory and anti-nociceptive drugs
Berger WT, et al.
PLoS ONE, 7(12) (2012)
Tracking of drug release and material fate for naturally derived omega-3 fatty acid biomaterials
Faucher KM, et al.
Annals of Biomedical Engineering, 44(3), 782-792 (2016)
E J Murphy
The American journal of physiology, 275(2 Pt 1), G237-G243 (1998-08-04)
The effects of sterol carrier protein-2 (SCP-2) expression on fatty acid uptake and cytoplasmic diffusion were determined using L cell fibroblasts transfected with cDNA encoding either the 15- or 13. 2-kDa form of SCP-2. Cis-parinarate and 12-N-methyl-(7-nitrobenz-2-oxa-1,3-diazol)aminostearate (NBD-stearate) were used
Targeting fatty acid binding protein (FABP) anandamide transporters-a novel strategy for development of anti-inflammatory and anti-nociceptive drugs
Berger WT, et al.
Testing, 7(12) (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service