Skip to Content
Merck
All Photos(1)

Key Documents

791642C

Avanti

15:0-18:1-d7-PA

Avanti Research - A Croda Brand

Synonym(s):

1-pentadecanoyl-2-oleoyl(d7)-sn-glycero-3-phosphate (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€230.00

€230.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€230.00

About This Item

Empirical Formula (Hill Notation):
C36H61D7NaO8P
Molecular Weight:
689.93
UNSPSC Code:
12352211
NACRES:
NA.25

€230.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (791642C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL (791642C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

15:0-18:1-d7-PA (1-pentadecanoyl-2-oleoyl(d7)-sn-glycero-3-phosphate) is a deuterated phospholipid. The lipid comprises phosphoglycerol attached to pentadecanoic acid and oleic acid. Oleic acid harbors the deuterium ions.
This deuterium labeled lipid is also available in a formulation of multiple lipid standards. The SPLASH LIPIDOMIX Standard includes all of the major lipid classes at ratios relative to human plasma, allowing one easy internal standard solution to be added to every sample.

Application

15:0-18:1-d7-PA (1-pentadecanoyl-2-oleoyl(d7)-sn-glycero-3-phosphate (sodium salt) has been used:
  • as a lipid internal standard in liquid chromatography-electrospray ionization quadrupole time-of-flight mass spectrometry (LC-ESI-QToF-MS) analysis for the extraction and profiling of lipids from egg yolk samples[1]
  • as an internal standard for yeast lipid quantification using nanoelectrospray ionization mass spectrometry[2]
  • as an isotopically labeled lipid internal standard for correction of lipids detected in lipid droplets using shotgun mass spectrometry analysis[3]

Packaging

2 mL Amber Glass Sealed Ampule (791642C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
LIPIDOMIX is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fenja Odendall et al.
Molecular biology of the cell, 30(21), 2681-2694 (2019-09-05)
Mitochondria are unique organelles harboring two distinct membranes, the mitochondrial inner and outer membrane (MIM and MOM, respectively). Mitochondria comprise only a subset of metabolic pathways for the synthesis of membrane lipids; therefore most lipid species and their precursors have
Ya Xie et al.
Food chemistry, 319, 126547-126547 (2020-03-18)
In this research, a novel ultrasound-assisted extraction (USAE) method using a one-phase solvent (isopropanol, IPA) combined with liquid chromatography-electrospray ionization quadrupole time-of-flight mass spectrometry (LC-ESI-QToF-MS) was developed for exhaustive profiling of egg yolk lipids, which are considered to be essential
Tong Liu et al.
Expert review of proteomics, 17(3), 233-242 (2020-04-21)
Background: Colorectal cancer (CRC) represents a third leading cause of cancer-related death worldwide. The reliable diagnostic biomarkers for detecting CRC at early stage is critical for decreasing the mortality.Method: A conjunctive lipidomic approach was employed to investigate the differences in
Florian Geltinger et al.
GeroScience, 42(1), 19-38 (2019-11-05)
Originally Lipid droplets (LDs) were considered as being droplets for lipid storage only. Increasing evidence, however, demonstrates that LDs fulfill a pleiotropy of additional functions. Among them is the modulation of protein as well as lipid homeostasis. Under unfavorable pro-oxidative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service