Skip to Content
Merck
All Photos(1)

Key Documents

791274P

Avanti

16:0 PE-DTPA (Gd)

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-diethylenetriaminepentaacetic acid (gadolinium salt), powder

Synonym(s):

1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine-N-diethylenetriaminepentaacetic acid (gadolinium salt); DPPE-DTPA (Gd)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€163.00

€163.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€163.00

About This Item

Empirical Formula (Hill Notation):
C51H98GdN6O17P
CAS Number:
Molecular Weight:
1255.58
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.23

€163.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (791274P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 791274P

shipped in

dry ice

storage temp.

−20°C

Application

16:0 PE-DTPA (Gd) may be used in the preparation of gadolinium-diethylenetriaminepentaacetic (Gd-DTPA)-loaded mannosylated liposomes.[1]

Biochem/physiol Actions

1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-diethylenetriaminepentaacetic acid (PE-DTPA) along with Gd-DTPA, an imaging reagent exhibit high molecular relaxivity and enhance spin-lattice" relaxation time (T1WI) contrast for magnetic resonance imaging (MRI).[1]

Packaging

5 mL Amber Glass Screw Cap Vial (791274P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mannose-coated gadolinium liposomes for improved magnetic resonance imaging in acute pancreatitis
Tian B, et al.
International journal of nanomedicine, 12, 1127-1127 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service