Skip to Content
Merck
All Photos(1)

Key Documents

790427P

Avanti

06:0 PC-d22

Avanti Research - A Croda Brand 790427P, powder

Synonym(s):

1,2-dihexanoyl-d22-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€163.00

€163.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€163.00

About This Item

Empirical Formula (Hill Notation):
C20H18NO8PD22
CAS Number:
Molecular Weight:
475.64
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€163.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (790427P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 790427P

shipped in

dry ice

storage temp.

−20°C

General description

1,2-dihexanoyl-sn-glycero-3-phosphocholine (06:0 PC) is a hydrophobic acyl chain containing phospholipid with a critical micelle concentration of 15 mM.[1] 06:0 PC-d22 is the deuterated form of 06:0 PC.
Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.

Application

06:0 PC-d22 has been used in small isotropic bicelles preparation for C-peptide membrane interaction studies by diffusion nuclear magnetic resonance (NMR) and 2D total correlation spectroscopy (TOCSY)[2] and in in situ infrared (IR) absorption spectra.[3] It may be used as an internal standard in capillary electrophoresis time of flight mass spectrometer (CE-TOFMS) and liquid chromatography with tandem mass spectrometry (LC-MS-MS) for the analysis of algal lipidome.[4]

Biochem/physiol Actions

1,2-dihexanoyl-sn-glycero-3-phosphocholine (06:0 PC) bicelle with 1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC) takes a disc like conformation.[5] Phosphocholine (PC) is the catabolized product of phosphatidylcholine (lecithin). PC mediates immune response and acts as a ligand for nicotinic acetylcholine receptors (nAChR).[6]

Packaging

5 mL Clear Glass Sealed Ampule (790427P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sofia Unnerståle et al.
Journal of biophysics (Hindawi Publishing Corporation : Online), 2012, 185907-185907 (2012-08-01)
C-peptide is the connecting peptide between the A and B chains of insulin in proinsulin. In this paper, we investigate the interaction between C-peptide and phospholipid bicelles, by circular dichroism and nuclear magnetic resonance spectroscopy, and in particular the pH
Takuro Ito et al.
Metabolomics : Official journal of the Metabolomic Society, 9(Suppl 1), 178-187 (2013-03-07)
Oil-rich algae have promising potential for a next-generation biofuel feedstock. Pseudochoricystis ellipsoidea MBIC 11204, a novel unicellular green algal strain, accumulates a large amount of oil (lipids) in nitrogen-deficient (-N) conditions. Although the oil bodies are easily visualized by lipophilic
Soichiro Matsunaga et al.
Langmuir : the ACS journal of surfaces and colloids, 33(46), 13157-13167 (2017-08-02)
In situ dynamic observation of model biological cell membranes, formed on a water/gold substrate interface, has been performed by the combination of electrochemical scanning tunneling microscopy and reflection infrared absorption vibrational spectroscopy. Monolayers of 1,2-dihexanoyl-sn-glycero-3-phosphocholine (DHPC) were formed on alkanethiol-modified
K Richter et al.
Scientific reports, 6, 28660-28660 (2016-06-29)
We demonstrated previously that phosphocholine and phosphocholine-modified macromolecules efficiently inhibit ATP-dependent release of interleukin-1β from human and murine monocytes by a mechanism involving nicotinic acetylcholine receptors (nAChR). Interleukin-1β is a potent pro-inflammatory cytokine of innate immunity that plays pivotal roles
Johannes Björnerås et al.
Biochimica et biophysica acta, 1848(11 Pt A), 2910-2917 (2015-09-06)
Mixtures of lipids and detergents are known to form bicelles at certain parameter ranges, but many uncertainties remain concerning the details of the phase behaviour of these mixtures and the morphology of the formed lipid assemblies. Here we used nuclear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service