Skip to Content
Merck
All Photos(1)

Key Documents

700199P

Avanti

Ursodeoxycholic acid

Avanti Research - A Croda Brand

Synonym(s):

5β-cholanic acid-3α,7β-diol

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€178.00

€178.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€178.00

About This Item

Empirical Formula (Hill Notation):
C24H40O4
CAS Number:
Molecular Weight:
392.57
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€178.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

package of 1 × 10 mg

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

bile acids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@]12[C@]([C@](CC[C@@H](O)C3)(C)[C@]3([H])C[C@@H]2O)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCC(O)=O

InChI key

RUDATBOHQWOJDD-UZVSRGJWSA-N

Packaging

5 mL Amber Glass Screw Cap Vial (700199p-10MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service