Skip to Content
Merck
All Photos(1)

Key Documents

700136P

Avanti

7α,27-dihydroxycholesterol-d6

Avanti Research - A Croda Brand 700136P, powder

Synonym(s):

7α,27-dihydroxycholesterol-d6

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€802.00

€802.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€802.00

About This Item

Empirical Formula (Hill Notation):
C27H40D6O3
CAS Number:
Molecular Weight:
424.69
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€802.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700136P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700136P

shipped in

dry ice

storage temp.

−20°C

General description

7α,27-dihydroxycholesterol (7α,27-di-OHC) is synthesized by the hydroxylation of 27-hydroxycholesterol (27-OHC).[1] It is an oxysterol with side chains oxidized[2] and is a structural isomer of 7α,25-dihydroxycholesterol.[1] 7α,27-dihydroxycholesterol-d6 is a deuterated derivative of 7α,27-di-OHC.

Application

7α,27-dihydroxycholesterol-d6 has been used as a deuterated internal standard in liquid chromatography electrospray ionization tandem mass spectrometric (LC/ESI-MS/MS) for human plasma oxysterol analysis.[1]

Biochem/physiol Actions

7α,27-dihydroxycholesterol (7α,27-di-OHC) is a ligand for Epstein-Barr virus-induced gene 2 (EBI2).[1] 7α27-di-OHC is implicated in neurological disease, Smith-Lemli-Opitz syndrome, obesity metabolic syndrome and diabetes by lipidomics analysis of human cerebrospinal fluid (CSF) samples.[3] The levels of 7α,27-di-OHC decreases in lipopolysaccharide activation.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (700136P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara Sottero et al.
Free radical biology & medicine, 144, 55-71 (2019-05-30)
High amounts of cholesterol have been definitely associated with the pathogenesis of several diseases, including metabolic and neurodegenerative disorders, cardiovascular diseases, and cancer. In all these pathologies the exacerbation of pro-oxidant and inflammatory responses is a consistent feature. In this
Valentin Mutemberezi et al.
Journal of neuroinflammation, 15(1), 74-74 (2018-03-11)
Oxysterols are cholesterol derivatives that have been suggested to play a role in inflammatory diseases such as obesity, atherosclerosis, or neuroinflammatory diseases. However, the effect of neuroinflammation on oxysterol levels has only been partially studied so far. We used an
Katharina R Beck et al.
Journal of lipid research, 60(9), 1535-1546 (2019-07-06)
Oxysterols previously were considered intermediates of bile acid and steroid hormone biosynthetic pathways. However, recent research has emphasized the roles of oxysterols in essential physiologic processes and in various diseases. Despite these discoveries, the metabolic pathways leading to the different
Ratna Karuna et al.
Steroids, 99(Pt B), 131-138 (2015-02-17)
We report a straightforward sample preparation procedure and a direct liquid chromatography electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS) method for the analysis of 7alpha,25-dihydroxycholesterol (7α25-OHC) and 7alpha,27-dihydroxycholesterol (7α27-OHC). By applying a slow protein precipitation approach using cold ethanol, we were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service