Skip to Content
Merck
All Photos(1)

Key Documents

700120P

Avanti

7α,24(R/S)-dihydroxycholestenone-d7

7α,24(R/S)-dihydroxycholestenone-d7, powder

Synonym(s):

111114

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€764.00

€764.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€764.00

About This Item

Empirical Formula (Hill Notation):
C27H37D7O3
CAS Number:
Molecular Weight:
423.68
MDL number:
UNSPSC Code:
41141804
NACRES:
NA.25

€764.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700120P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700120P

shipped in

dry ice

storage temp.

−20°C

General description

7α,24(R/S)-dihydroxycholestenone-d7 is a deuterated derivative of the oxysterol 7α,24(R/S)-dihydroxycholestenone. 7α,24-dihydroxy-3-oxocholest-4-en-26-oic acid is synthesized from 3β,7α,24 S-trihydroxycholest-5-en-(25 R)26-oic acid ((25 R)CA5-3β,7α,24 S-triol) in the presence of enzyme 3β-hydroxysteroid dehydrogenase type 7 (HSD3B7). 7α,24(S)-dihydroxycholestenone is also derived by the 7α-hydroxylation of 24S hydroxycholesterol (24S-HC) in the presence of enzyme oxysterol 7-α-hydroxylase 2 (CYP39A1).[1][2]

Packaging

5 mL Amber Glass Screw Cap Vial (700120P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxysterols as lipid mediators: Their biosynthetic genes, enzymes and metabolites
Griffiths WJ and Wang Y
Prostaglandins & other lipid mediators null
Concentrations of bile acid precursors in cerebrospinal fluid of Alzheimer's disease patients
Griffiths WJ, et al.
Free radical biology & medicine null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service