Skip to Content
Merck
All Photos(2)

Key Documents

700083P

Avanti

3-oxo-4-cholestenoic acid

Avanti Research - A Croda Brand

Synonym(s):

cholest-4-en-3-one-27-oic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€403.00

€403.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€403.00

About This Item

Empirical Formula (Hill Notation):
C27H42O3
CAS Number:
Molecular Weight:
414.62
UNSPSC Code:
12352211
NACRES:
NA.25

€403.00


Please contact Customer Service for Availability

Request a Bulk Order

description

(25R)-cholest-4-en-26-oic acid, 3-oxo

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700083P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC1(CC2)C(CCC3C1CCC4(C)C3CCC4C(CCCC(C)C(O)=O)C)=CC2=O

InChI key

PSXQJZDFWDKBIP-UHFFFAOYSA-N

General description

3-oxo-4-cholestenoic acid (3OChA) is synthesized from cholesterol by the oxidation at the positions C3 and C26 in the presence of P450 cytochromes (CYP125 and CYP142). This pathway is well established in M. smegmatis.[1] 3OChA is an intermediate in cholesterol catabolism and is more soluble compared to cholesterol.[1]

Application

3-oxo-4-cholestenoic acid has been used as a inducer ligand for the TetR-like repressor (KstR) of M. smegmatis.[1]

Biochem/physiol Actions

3-oxo-4-cholestenoic acid (3OChA) acts as an inducer to the TetR-like repressor (KstR) and modulates the expression kstR regulon genes.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (700083P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esther García-Fernández et al.
The Journal of biological chemistry, 289(25), 17576-17588 (2014-05-08)
Cholesterol degradation plays a prominent role in Mycobacterium tuberculosis infection; therefore, to develop new tools to combat this disease, we need to decipher the components comprising and regulating the corresponding pathway. A TetR-like repressor (KstR) regulates the upper part of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service