Skip to Content
Merck
All Photos(2)

Key Documents

700042P

Avanti

4β-hydroxycholesterol-d7

Avanti Research - A Croda Brand

Synonym(s):

cholest-5-ene-3b,4b-diol(d7)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€382.00

€382.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€382.00

About This Item

Empirical Formula (Hill Notation):
C27H39O2D7
CAS Number:
Molecular Weight:
409.70
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

€382.00


Please contact Customer Service for Availability

Request a Bulk Order

description

cholest-5-ene-3,4-diol-d7

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700042P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](C2=CC[C@H]3[C@H]4[C@@]([C@H](CC4)[C@@H](CCCC(C([2H])([2H])[2H])(C([2H])([2H])[2H])[2H])C)(CC[C@@H]3[C@]2(CC1)C)C)O

InChI

1S/C27H46O2/c1-17(2)7-6-8-18(3)20-11-12-21-19-9-10-23-25(29)24(28)14-16-27(23,5)22(19)13-15-26(20,21)4/h10,17-22,24-25,28-29H,6-9,11-16H2,1-5H3/t18-,19+,20-,21+,22+,24+,25-,26-,27-/m1/s1/i1D3,2D3,17D

InChI key

CZDKQKOAHAICSF-BZNWTYAPSA-N

General description

4β-hydroxycholesterol (4β-HC) is a catabolic product of cholesterol metabolism mediated by the enzyme cytochrome P450 family 3 subfamily A member 4 (CYP3A4).[1] 4β-hydroxycholesterol-d7 is the deuterated form of 4β-hydroxycholesterol.

Application

4β-hydroxycholesterol-d7 has been used:
  • as an internal standard to spike plasma samples ultra-high-pressure liquid,chromatography–tandem mass spectrometry (UHPLC-MS/MS) analysis[1]
  • to spike serum samples for mass spectrometry measurements[2]
  • as an internal standard for the quantification of human colon biopsies samples using HPLC-MS[3]

Biochem/physiol Actions

4β-hydroxycholesterol (4β-HC) may function as a liver X receptor ligand.[3] The levels of 4β-HC are elevated in dextran sulfate induced colitis along with other oxysterols and it may promote colon inflammation.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (700042P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Owein Guillemot-Legris et al.
Journal of Crohn's & colitis, 13(2), 218-229 (2018-10-09)
Inflammatory bowel diseases [IBD] represent a challenging health issue with a complex aetiology involving genetic and environmental parameters. Although our understanding of the pathophysiology of IBD has improved, much remains to be explored. In this context, bioactive lipids, more specifically
Mahmoud Hasan et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1033-1034, 193-199 (2016-08-28)
Cytochrome P450 3A (CYP) enzymes are involved in the elimination of many drugs and are known to be regulated by several environmental factors. Thus, it was the aim of this study to develop and validate an analytical method allowing estimation
Sarah J Woolsey et al.
Basic & clinical pharmacology & toxicology, 118(4), 284-291 (2015-09-25)
Due to high basal interindividual variation in cytochrome P450 3A (CYP3A) activity and susceptibility to drug interactions, there has been interest in the application of efficient probe drug phenotyping strategies, as well as endogenous biomarkers for assessment of in vivo

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service