Skip to Content
Merck
All Photos(1)

Key Documents

330001S

Avanti

E06 mAb

E06 Mouse Monoclonal Antibody (IgM), Anti-(Oxidized Phospholipid), 1X PBS (Phosphate Buffered Saline) with 0.27 mM Na2 EDTA

Synonym(s):

E06 mAb, T15 antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
application:
citations:
9

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

Assay

>95%

form

solution

packaging

pkg of 1 × 0.1 mL (polypropylene tube with screw cap (330001S-100ug))

manufacturer/tradename

Avanti Research - A Croda Brand 330001S

concentration

1 mg/mL (330001S-100ug)

shipped in

dry ice

storage temp.

−70°C

General description

The E06 antibody is a murine monoclonal IgM natural antibody that binds to oxidized phospholipid and reacts with oxidized LDL, oxidized HDL and protein covalently modified by oxidized phospholipid. Each batch is verified for activity and provided with detailed protocols for the above applications.The E06 antibody was developed by Dr. Joseph Witztum at the University of California, San Diego using apolipoprotein E knock-out mice fed a high-fat diet for seven months.

Application

E06 mAb has been used as a marker to detect oxidative injury.[1]
The E06 antibody has been used to probe atherosclerotic plaque in mice, zebrafish, rabbits, monkeys and humans. It also binds to apoptotic cells, but not viable cells, and can be used in a variety of applications, including ELISA, immunohistochemistry and Western blot analysis, as well as for in vivo imaging with MRI techniques.

Packaging

0.5 mL Polypropylene Tube with Screw Cap (330001S-100ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Friedman et al.
The Journal of biological chemistry, 277(9), 7010-7020 (2001-12-18)
The oxidation of low density lipoproteins (LDL) has been correlated with atherogenesis through a variety of pathways. The process involves nonspecific fragmentation, oxidative breakdown, and modification of the lipids and protein of LDL. The process yields a variety of bioactive
J Qin et al.
Journal of neuroscience research, 85(5), 977-984 (2007-02-17)
Multiple sclerosis (MS) is a common autoimmune neurodegenerative disease of unknown cause, which results in inflammation and plaques of demyelination in brain and eventual axonal degeneration. We report the novel presence of oxidized phosphatidylcholine [1-palmitoyl-2-(5'-oxo)valeryl-sn-glycero-3-phosphorylcholine (POVPC)], a lipid associated with
W Palinski et al.
The Journal of clinical investigation, 98(3), 800-814 (1996-08-01)
Many reactive products may be formed when LDL undergoes lipid peroxidation, which in turn can react with lipids, apoproteins, and proteins, generating immunogenic neoepitopes. Autoantibodies recognizing model epitopes of oxidized low density lipoprotein, such as malondialdehydelysine, occur in plasma and
Arnab Chattopadhyay et al.
Journal of lipid research, 57(5), 832-847 (2016-03-12)
Mouse chow supplemented with lysophosphatidylcholine with oleic acid at sn-1 and a hydroxyl group at sn-2 (LysoPC 18:1) increased LysoPC 18:1 in tissue of the jejunum of LDL receptor (LDLR)-null mice by 8.9 ± 1.7-fold compared with chow alone. Western
Oxidative tissue injury in multiple sclerosis is only partly reflected in experimental disease models
Schuh C, et al.
Acta Neuropathologica, 128(2), 247-266 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service