Skip to Content
Merck
All Photos(1)

Documents

181108C

Avanti

Liver Extract Polar

Avanti Research - A Croda Brand

Synonym(s):

Liver Polar Lipid Extract (Bovine)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

chloroform solution

packaging

pkg of 2 × 20 mL (181108C-1g)
pkg of 1 × 4 mL (181108C-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

25 mg/mL (181108C-100mg)
25 mg/mL (181108C-1g)

lipid type

lipid extracts

shipped in

dry ice

storage temp.

−20°C

General description

Polar Lipid Extract is derived from the total lipid extract by precipitation with acetone followed by extraction of the acetone insoluble material with diethyl ether.

Application

Liver Extract Polar has been used to measure the biochemical components of non-tumourigenic rat fibroblast cell line, M1, and its highly tumourigenic counterpart, MR1. It has also been used in the preparation of lipid and bile acid solutions.

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (181108C-1g)
5 mL Clear Glass Sealed Ampule (181108C-100mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Bile acids stimulate ATP hydrolysis in the purified cholesterol transporter ABCG5/G8
Johnson B J H, et al.
Biochemistry, 49(16), 3403-3411 (2010)
Methods for measuring the infrared spectra of biological cells
Mourant J R, et al.
Physics in Medicine and Biology, 48(2), 243-243 (2003)
Wenbo Cao et al.
Nature communications, 11(1), 375-375 (2020-01-19)
Lipids play a pivotal role in biological processes and lipid analysis by mass spectrometry (MS) has significantly advanced lipidomic studies. While the structure specificity of lipid analysis proves to be critical for studying the biological functions of lipids, current mainstream
Wenpeng Zhang et al.
Analytical chemistry, 92(9), 6719-6726 (2020-04-10)
Mapping the complete molecular composition of a lipidome is considered an important goal of lipidomics for unraveling pathways and mechanisms behind lipid homeostasis. Conventional dissociation methods of mass spectrometry (MS) usually cannot give detailed structural information on lipids such as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service