Skip to Content
Merck
All Photos(1)

Key Documents

ATEH94724FBE

AstaTech, Inc.

4-BROMO-3-FLUORO-N,N-DIMETHYLBENZAMIDE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
246.08
MDL number:

Iupac Name

4-bromo-3-fluoro-N,N-dimethylbenzamide

Inchi Code

1S/C9H9BrFNO/c1-12(2)9(13)6-3-4-7(10)8(11)5-6/h3-5H,1-2H3

InChI key

SQSHZRBCVHCKPX-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=173837

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=C91410


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library