AOBH945512CD
AOBChem
2-bromo-1-chloro-4-(cyclohexylmethyl)benzene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2-bromo-1-chloro-4-(cyclohexylmethyl)benzene
Inchi Code
1S/C13H16BrCl/c14-12-9-11(6-7-13(12)15)8-10-4-2-1-3-5-10/h6-7,9-10H,1-5,8H2
InChI key
AYJQVVMPCRFDHX-UHFFFAOYSA-N
Purity
97
Country of Origin
CN
Physical Form
Liquid
Product Link
https://aobchem.com/2-bromo-1-chloro-4-cyclohexylmethyl-benzene.html
MSDS
https://aobchem.com/pub/media/files/sds/47245.pdf
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service