Iupac Name
3-bromo-5-chloro-1,1'-biphenyl
Inchi Code
1S/C12H8BrCl/c13-11-6-10(7-12(14)8-11)9-4-2-1-3-5-9/h1-8H
InChI key
SJLJTKRBBCLGPR-UHFFFAOYSA-N
storage temp.
2-8C
Purity
95
Country of Origin
CN
Shipping Temperature
Room Temperature
Physical Form
Liquid
Product Link
https://aobchem.com/3-bromo-5-chlorobiphenyl.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://aobchem.com/pub/media/files/sds/27446.pdf
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service