Skip to Content
Merck
All Photos(1)

Key Documents

AOB640091409

AOBChem

4-Bromoacetyl-3-fluorophenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
260.86
MDL number:

Iupac Name

4-(bromoacetyl)-3-fluorophenylboronic acid

Inchi Code

1S/C8H7BBrFO3/c10-4-8(12)6-2-1-5(9(13)14)3-7(6)11/h1-3,13-14H,4H2

InChI key

BXVHKBNJUHSCRF-UHFFFAOYSA-N

Purity

97

Country of Origin

CN

Product Link

https://aobchem.com/4-bromoacetyl-3-fluorophenylboronic-acid.html

MSDS

https://aobchem.com/pub/media/files/sds/10354.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library