Z764736
Ultrasonic processors 500W and 750W
750W, AC/DC input 220 V AC, UK plug
Synonym(s):
Ultrasonic Processor
Select a Size
€6,480.00
Select a Size
About This Item
€6,480.00
Recommended Products
feature
CE compliant
manufacturer/tradename
Sonics & Materials VCX-750UK-220V
Looking for similar products? Visit Product Comparison Guide
General description
- Automatic tuning and frequency control
- Integrated temperature controller from 1°C to 100°C
- Stores up to 10 procedures
- Real time display
- Control process time from 1 second to 10 hours
- Shipped complete with a 1/2 in. (13 mm) probe (630-0220) with replaceable tip, tool kit, and instruction manual
Applications: Cell disruption in bacteria, spores, and tissues; accelerating catalytic reactions; extractions of serums, toxins, enzymes, and viruses from organic sources; solubilizing difficult compounds; emulsification to 0.01 mm; homogenizing liquids; sample preparation from microscopy and particle size analysis.
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service