Key Documents
Z756997
Kern DBS moisture analyzer, for samples up to 60 g
AC/DC input 110 V AC
Synonym(s):
moisture sensor, moisture tester
About This Item
UNSPSC Code:
41111502
Pricing and availability is not currently available.
manufacturer/tradename
Kern DBS 60-3 US
weighing capacity measuring range
60 g
L × W × H
202 mm × 336 mm × 157 mm
General description
  • Halogen quartz glass heater
  • Observation window over the sample
  • Internal memory for automatic sequence of 10 drying programs
  • Sample description for up to 99 samples
  • Date and time display as standard
  • 10 sample plates included
Choose from one of the most recent versions:
Certificates of Analysis (COA)