Key Documents
Z744084
Sigma-Aldrich
Water Purifier Cartridge for H-Genie® Lite
Select a Size
1 PKG
€1,570.00
€1,570.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
€1,570.00
About This Item
UNSPSC Code:
12161503
NACRES:
NA.22
€1,570.00
Please contact Customer Service for Availability
Request a Bulk Order
General description
Water Purifier Cartridge for H-Genie® Lite is designed to keep the water purity at the required level to maintain optimum cell lifetime. Expected consumption with normal daily use is 6 cartridges per a year.
H-Genie® Lite water purifier cartridges are to be used with H-Genie® Lite benchtop hydrogen generator (Z744083)
Legal Information
H-Genie is a registered trademark of ThalesNano Energy
Choose from one of the most recent versions:
Certificates of Analysis (COA)