Key Documents
Z742598
Accuris SmartDoc narrow band pass filter
Orange PMMA; Orange photo filter; Used for imaging green stains
Synonym(s):
Accuris SmartDoc
Select a Size
1 EA
€34.50
€34.50
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€34.50
About This Item
UNSPSC Code:
41106000
NACRES:
NB.79
€34.50
Please contact Customer Service for Availability
feature
Orange PMMA; Orange photo filter; Used for imaging green stains
packaging
pkg of 1
manufacturer/tradename
Benchmark Scientific E5001-ORANGE
General description
An orange photo filter used to image green stains on blue light illuminators and transilluminators.
Legal Information
Accuris is a trademark of Benchmark Scientific, Inc.
SmartDoc is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)