Key Documents
Z742353
2MAG Stirring bar remover XL
with integrated corrosion-resistant high-energy magnet
Synonym(s):
magnetic extraction tool
Select a Size
1 EA
€431.00
€431.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€431.00
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
€431.00
Please contact Customer Service for Availability
manufacturer/tradename
2mag 45100
General description
Extremely strong stirring bar remover with an integrated corrosion-resistant high energy magnet, ideal for reliable inserting and removing of strong and heavy magnetic stirring bars with a weight up to 1,000 g. Allows for simple handling due to a special developed stripping mechanism, constructed from stainless steel. Solid and easy to clean with high chemical resistance. Length of 100 cm for use in deep reaction vessels.
Choose from one of the most recent versions:
Certificates of Analysis (COA)