Skip to Content
Merck
All Photos(7)

Key Documents

Z742108

Sigma-Aldrich

KitAlysis 24-well Reaction Block and Inertion Box Combination Starter Set

Synonym(s):

Benchtop inertion box

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€1,970.00

€1,970.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€1,970.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.51

€1,970.00


Please contact Customer Service for Availability

Request a Bulk Order

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling

Quality Level

General description

The perfect KitAlysis Labware starter kit that combines the KitAlysis Inertion Box (Z742064) with the KitAlysis 24-Well Reaction Block and Screwdriver Set (Z742107).

Provides an inert environment to run oxygen sensitive cross-coupling reactions in a laboratory fume hood.
Designed to be used with KitAlysis High-Throughput Screening Kits.

Components:
  • Ιnertion Box
  • 24-Well Reaction Block
  • Torque Screwdriver set with bit
  • Reaction Block Replacement Screws (10ea)

Features and Benefits

Inertion Box
  • Run 24 micro-scale reactions in parallel under an inert atmosphere.
  • Provides a constant flow of inert gas over working area.
  • Easy attachment of tubing (not included) from inert gas valve on hood or stand-alone tank

24-well Reaction Block and Screwdriver Set
  • Torque Screwdriver specially designed to seal the block for minimal solvent loss while heating at elevated temperatures.
  • Torque setting prevents over-tightening and stripping of screws.
  • Includes small screwdriver to easily remove torqued screws after reaction is complete.

Legal Information

KitAlysis is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

KitAlysis High-Throughput Screening Kits provide solution to efficiently identify or optimize suitable catalytic reaction conditions. Chemist can rapidly run 24 unique micro scale reactions in parallel with tailored conditions.

Tools aid in kit setup for organic chemistry techniques, ensuring ease and success.

PhotoKitAlysis Reaction Kit aids in cross-coupling reactions, offering pre-weighed components for optimization.

Materials Included in your KITALYSIS-24PD-2PK High-Throughput Screening Kit

Protocols

Solutions & slurries to make: Go to the online user set-up page to enter molecular weights into the downloadable excel file.

A step-by-step protocol guide for KitAlysis Suzuki-Miyaura Cross-Coupling Reaction Screening Kit.

Catalyst screening kits, KitAlysis™, and TLC-MS analysis enable high-throughput synthesis and analysis for efficient reactions.

See All

Related Content

Solutions & slurries to make: Go to the online user set-up page to enter molecular weights into the downloadable excel file.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service