Z740892
Aldrich® Essentials graduated cylinder, class B, meets ASTM E 1272
capacity 1000 mL, subdivision, 10.0 mL, tol. 10.0 mL, Graduated "To Contain", plastic bumper, hex base
About This Item
Recommended Products
feature
subdivision 10.0 mL
packaging
pkg of 4 ea
tol. measuring range
10.0 mL
capacity
1000 mL
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service