Key Documents
Z687693
eVol® electronic syringe NMR starter kit
CE compliant
About This Item
UNSPSC Code:
41121705
NACRES:
SB.32
Pricing and availability is not currently available.
feature
CE compliant
manufacturer/tradename
SGE 2910100
General description
"Enables Precise, Intuitive, Operator-independent Dispensing"
eVol® NMR Edition allows accurate manipulation of small volumes of NMR samples enabling dilution directly in the analysis tube - even in tubes with the smallest internal diameters.
eVol facilitates in-tube sample mixing and enables recovery of precious samples allowing them to be stored in alternative vessels (such as inexpensive vials) rather than being left in valuable tubes. These tubes can now be emptied using eVol, for washing and re-use.
The First High-Tech Electronic Syringe for NMR Capillaries
eVol is the coupling of two precision devices: a digitally controlled electronic drive and an XCHANGE® (patent pending) enabled analytical syringe. The result is a digitally controlled positive displacement dispensing system that can be programmed to reproducibly and accurately perform a wide variety of liquid handling procedures.
  • XCHANGE analytical syringes are easily and quickly changed allowing them to be dedicated to individual liquids, and methods, to prevent possible cross-contamination of reagents
  • Unlike air-displacement devices, eVol is the perfect solution for accurately aspirating and dispensing both aqueous and non-aqueous liquids
  • eVol features a familiar touch wheel operator interface and a full-color screen. The clever menu allows all the functions to be accessed logically and quickly. The programming functions are intuitive and include help screens and prompts
  • eVol is ergonomic, comfortable, and intuitive to operate
World′s First Operator Calibrated Analytical Syringe
  • To comply with stringent laboratory standards (e.g. GLP, GMP, FDA) eVol can be easily calibrated to ensure accurate dispensing at all times. The gravimetric calibration procedure is simple and can be performed at operator defined intervals
  • Calibration factors can be stored for up to ten XCHANGE syringes and can be quickly loaded when the syringe is changed
The eVol NMR kit contains the following:
  • eVol - Automated Analytical Syringe
  • Manual, Quick-Start Guide & Support CD
  • Multi-Country Charger
  • Storage Stand
  • 5 μL eVol XCHANGE syringe with 11.5 cm needle
  • 50 μL eVol XCHANGE syringe with 11.5 cm and 18 cm needles
  • 500 μL eVol XCHANGE syringe with 11.5 cm and 18 cm needles
Legal Information
XCHANGE is a registered trademark of SGE Analytical Science Pty Ltd.
eVol is a registered trademark of SGE Analytical Science Pty Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)