Skip to Content
Merck
All Photos(1)

Key Documents

Z678481

Krytox® XP vacuum oil

grade, 1514

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
40161501
NACRES:
NB.79

mol wt

2840

feature

grade 1514

manufacturer/tradename

Du Pont

viscosity number

(20 °C) (142 mm2/s)

density

1.89 g/mL at 20 °C

Looking for similar products? Visit Product Comparison Guide

General description

  • Rust preventing
  • Nonflammable
  • Nonreactive
  • Free from acetal groups
  • Recyclable
Krytox XP oils contain a soluble additive to prevent rust. This additive enhances the
performance of Krytox vacuum pump fluids, giving them improved performance
properties. The long-term antirust properties repel moisture, providing extra
protection from corrosion of metal parts and bearing surfaces.

  • Rust preventing
  • Nonflammable
  • Nonreactive
  • Free from acetal groups
  • Recyclable

Legal Information

Krytox is a registered trademark of E. I. du Pont de Nemours and Company

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service