Key Documents
Z675458
Sterile economy sample spoon
2.5 mL
Synonym(s):
dispensing tool
Select a Size
100 EA
€157.00
€157.00
Please contact Customer Service for Availability
Select a Size
Change View
100 EA
€157.00
About This Item
UNSPSC Code:
41122403
NACRES:
NB.35
€157.00
Please contact Customer Service for Availability
description
PS
manufacturer/tradename
Bel-Art F36740-00002
General description
Moulded of lightweight and rigid, white high impact polystyrene. Individually packaged sterile spoons for one time use packed 100 per box.
  • Use to collect specific volumes of dry or liquid materials
  • Remains upright when filled or empty for use as a weighing boat
Choose from one of the most recent versions:
Certificates of Analysis (COA)