Skip to Content
Merck
All Photos(1)

Documents

Z655570

Hanna combination ISE electrodes

Nitrate, Polymer membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115603

material

polyetherimide body

manufacturer/tradename

Hanna HI 4113

ISE measuring range

14000-0.1 ppm

pH measuring range

2.5-11

parameter

0-40 °C temp. range

electrode immersion depth × O.D.

120 mm × 12 mm

Looking for similar products? Visit Product Comparison Guide

General description

These include a sensor and reference electrode in a single electrode body. Hanna combination ion selective electrodes provide selectivity and include superior double junction reference into an electrode body. Combination solid-state electrodes have a built-in solid-state sensor and quick refill reference electrode.

Hanna′s liquid membrane and fluoride combination electrodes have replaceable module construction and the Hanna double junction refence stability.

Application

Determination of free nitrate in natural waters (fresh and sea), and in emulsified food and plant samples.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service