Key Documents
Z645826
Chemical spill cushions
size medium
Synonym(s):
chemical cleanup, spill cleanup, spill safety
Select a Size
20 EA
€146.00
€146.00
Please contact Customer Service for Availability
Select a Size
Change View
20 EA
€146.00
About This Item
UNSPSC Code:
47131905
NACRES:
NB.35
€146.00
Please contact Customer Service for Availability
size
medium
description
each cushion absorbs 4.5 Litres
manufacturer/tradename
Clear-spill CCM
availability
not available in ROW, USA and Canada
L × W
30 cm × 35 cm
General description
Cushions can be used with drip pans or in conjection with pads and socks for absorbing larger spills.
Choose from one of the most recent versions:
Certificates of Analysis (COA)