Key Documents
Z630020
IKA® HS 501 Horizontal shaker
AC/DC input 230 V AC
Synonym(s):
lab shaker, laboratory shaker, orbital shaker, reciprocating shaker
Select a Size
1 EA
€5,660.00
1 EA
€5,660.00
1 EA
€5,660.00
€5,660.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€5,660.00
1 EA
€5,660.00
1 EA
€5,660.00
About This Item
UNSPSC Code:
41103811
NACRES:
NB.73
€5,660.00
Please contact Customer Service for Availability
feature
CE compliant
packaging
pack of 1 ea
manufacturer/tradename
IKA 0025002982 (Europe) and 0025004443 (GB)
General description
comes with integrated USD and RS 232 interface
Low profile laboratory shaker with a pleasant design, large mounting surface and a loading capacity of 33 lbs., depending on the attachment. The horizontal shaker features a reciprocating motion. Infinitely variable speed control from 0-300 rpm, digitally displayed. Timer 0-56 min and setting for continuous operating. Ideally suitable for vessels with a volume of more than 250 mL, e.g. round flasks, Erlenmeyer flasks, culture flasks, culture bottles.
For biological and microbiological growth tests, for the production of stock solution, etc.
Guaranteed continuous operation even under extreme loads.
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)