Key Documents
Z569933
Sigma-Aldrich
Chem-FLEX transfer line with male and female Luer connectors
tube L 60 in.
Select a Size
1 EA
€109.00
€109.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€109.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
€109.00
Please contact Customer Service for Availability
tube L
60 in.
General description
One male and one female Luer lock connector with chrome-plated brass bodies are connected to Chem-Flex 106 tubing with nylon clamps. Chem-Flex tubing was developed by Aldrich for the safe transfer of aggressive solvents, air-sensitive liquids, and gases.
Chem-Flex Tubing Features:
Chemically inert, thin-wall PTFE inner tube bonded to a clear PVC outer tube for extra strong, flexible transfer lines that are resistant to crushing or kinking.
  • Luer fittings on ends of tubing allow easy connection to
  • Stopcocks and valves
  • Needles of all lengths and gauges
  • Syringes and pumps
Applications:
  • Pressure transfer of liquids- Using Luer needles, stopcocks
  • Reagent addition to reactions- Using Luer needle, stopcock, syringe or syringe pump
Legal Information
Chem-Flex is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)