Key Documents
Z563978
Büchi® heating bath for R-210/215 evaporators
AC/DC input 220 - 240 V AC
About This Item
UNSPSC Code:
41100000
Pricing and availability is not currently available.
description
Model B-491
manufacturer/tradename
Büchi 048200
parameter
20-180 °C temp. limit
capacity
≤4 L
General description
The heating bath, for water or oil covers a wide temperature range. A continuous display shows the actual temperature and the temperature setpoint. It has a cordless power supply so it is not necessary to unplug connections when filling or emptying the bath.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)