Skip to Content
Merck
All Photos(1)

Key Documents

Z549665

Sigma-Aldrich

Aldrich® stirring rods

diam. × H 10 mm × 400 mm

Sign Into View Organizational & Contract Pricing

Select a Size

12 EA
€144.00

€144.00


Please contact Customer Service for Availability


Select a Size

Change View
12 EA
€144.00

About This Item

UNSPSC Code:
41103800
NACRES:
NB.72

€144.00


Please contact Customer Service for Availability

packaging

pack of 12 ea

manufacturer/tradename

Sigma-Aldrich

diam. × H

10 mm × 400 mm

Looking for similar products? Visit Product Comparison Guide

General description

Features smooth, fire-polished ends for easy and safe handling.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service