Key Documents
Z549347
Sigma-Aldrich
Aldrich® anticlimb splash-guard adapter with fritted disc
female joint: ST/NS 29/32, male joint: ST/NS 24/40, capacity 250 mL
Select a Size
1 EA
€306.00
€306.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€306.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
€306.00
Please contact Customer Service for Availability
Joint
female joint: ST/NS29/32
male joint: ST/NS24/40
capacity
250 mL
pore size
70-100 μm porosity
Related Categories
General description
These traps greatly reduce carry-over from foaming or bumping. Because the venting hole is located at the top, the foam can be prevented from climbing higher into the condenser. The fritted disc prevents particulates from entering the condenser.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)