Documents
Z544523
Molymod® molecular model set
Student Organic/Inorganic
About This Item
UNSPSC Code:
60104506
NACRES:
NB.17
feature
no. of atoms 52
no. of bonds 38
manufacturer/tradename
Molymod MMS-009
General description
14 hydrogen (l), 6 carbon (lV), 7 oxygen (ll, lV), 3 nitrogen (lll, lV), 2 sulphur (ll, Vl), 2 phosphorus (lll, V), 6 halogen (l), 8 metal (l, ll, lll, lV, Vl), 4 brown (lll, lV, V, Vl), 3 lone pair 2D paddles (beige) Links: 20 medium (grey), 5 medium (purple), 12 long flexible (grey)
Top quality Molymod molecular model sets are for use by students, teachers, and researchers. May be upgraded or customized with additional model parts, for more info, go to sigma-aldrich.com and enter Molymod in our search engine.
Scale is 2.8 cm =100 pm (open style) and 1.45 cm = 100 pm (compact, semi-space-filling). Sets include standard color-coded plastic atom centers (with bond holes set at the appropriate angles), bonding links, and a plastic storage box.
Set pictured is representative of the Student set.
See our Selection Guide for detailed descriptions of set contents.
Legal Information
Molymod is a registered trademark of Spiring Enterprises Ltd.