Key Documents
Z530395
Sigma-Aldrich
Aldrich® short-path distillation head
thermometer joint: ST/NS 10/18, Joints: ST/NS 19/22 (2)
Select a Size
1 EA
€113.00
€113.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€113.00
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
€113.00
Please contact Customer Service for Availability
Joint
thermometer joint: ST/NS10/18
Joints: ST/NS19/22 (2)
immersion depth
50 mm
Related Categories
General description
Vigreux indents prevent bumping of fine or lightweight column packing into the condenser or fraction cutter when column is flooded.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)