Key Documents
Z508233
Sigma-Aldrich
Quick-release clamps
without extension, for flange 100 mm
Synonym(s):
clamps, quick release clamp
Select a Size
1 EA
€199.00
€199.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€199.00
About This Item
UNSPSC Code:
41122413
NACRES:
NB.43
€199.00
Please contact Customer Service for Availability
manufacturer/tradename
Ace Glass 651725
General description
SS clamps for use with reaction flasks and heads with a 100 mm Schott flange. The extension rod can be clamped to the support frame and helps stabilize the reaction assembly.
Choose from one of the most recent versions:
Certificates of Analysis (COA)