Key Documents
Z408433
Scienceware® ROTO-FLO flow indicator
fits tubing, 3/16 ‑ 1/4 in., blue wheel
Select a Size
1 EA
€34.00
€34.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€34.00
About This Item
UNSPSC Code:
20121904
NACRES:
NB.51
€34.00
Please contact Customer Service for Availability
description
straight tube ends
feature
fits tubing 3/16 ‑ 1/4 in.
manufacturer/tradename
Bel-Art
color
blue wheel
General description
Colored paddle wheel spins freely with liquid or gas flow (range: 0.5 to 10 L/min). Made of clear PS. Not for use with corrosive fluids nor at temperatures above 64 °C.
Legal Information
ROTO-FLO is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)